Benzyl alcohol, ACS, 99+%, Thermo Scientific Chemicals
Benzyl alcohol, ACS, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzyl alcohol, ACS, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
250 mL
1 L
4 x 1 L
Catalog number 041218.K7
also known as 041218-K7
Price (USD)/ Each
387.00
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
387.00
Add to cart
Benzyl alcohol, ACS, 99+%, Thermo Scientific Chemicals
Catalog number041218.K7
Price (USD)/ Each
387.00
-
Add to cart
Chemical Identifiers
CAS100-51-6
IUPAC Namephenylmethanol
Molecular FormulaC7H8O
InChI KeyWVDDGKGOMKODPV-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Color (APHA)20, max
Impurity content0.02%, max, as Acetophenone C₆H₅COCH₃ ( by GC)
Assay (GC)≥99.0%
Impurity content0.01%, max, as Benzaldehyde C₆H₅CHO ( by GC)
FormClear liquid
View more
Benzyl alcohol is widely used as a solvent for the dielectrophoretic reconfiguration of nanowires, inks, paints, lacquers and epoxy resin coatings and as a precursor to a variety of esters used in soaps, perfumes and flavoring. It is employed as a local anesthetic which reduces the pain associated with lidocaine injection. It has a various applications in baby products, bath products, soaps and detergents, eye makeup, blushers, cleansing products, make up products as well as hair, nail and skin care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only