Specifications
Chemical Name or MaterialCobalt(II) 2-methoxyethoxide
CAS142600-61-1
Health Hazard 1H226-H302+H312+H332-H314-H335-H360FD-H370-H373
Health Hazard 2GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more