Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight, Thermo Scientific Chemicals
Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 041238.14
also known as 041238-14
Price (USD)/ Each
36.50
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
36.50
Add to cart
Polyvinyl alcohol, 86-89% hydrolyzed, low molecular weight, Thermo Scientific Chemicals
Catalog number041238.14
Price (USD)/ Each
36.50
-
Add to cart
Chemical Identifiers
CAS9002-89-5
Molecular Formula(C2H4O)n
SMILESOC(-*)C-*
MDL NumberMFCD00081922
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay (unspecified)< 5% volatile content
FormGranular powder
Assay (unspecified)86.0 to 89.0 mole% hydrolyzed
Polyvinyl alcohol is used as a biological reagent to test the dispersion of zinc oxide nanoparticles. It is an effective protein resistant material and is used to stabilize gold colloids of diameter 3-8nm. It is used as an emulsion polymerization aid to make polyvinyl acetate, as a protective colloid, as a film used in the water transfer printing process, as a surfactant for the formation of polymer encapsulated nanobeads, as an embolization agent in medical procedures and as a fixative for specimen collection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only