Thermo Scientific Chemicals

Polyvinyl alcohol, 86-89% hydrolyzed, medium molecular weight, Thermo Scientific Chemicals

Catalog number: 041239.22
100 g, Each
Thermo Scientific Chemicals

Polyvinyl alcohol, 86-89% hydrolyzed, medium molecular weight, Thermo Scientific Chemicals

Catalog number: 041239.22
100 g, Each
Quantity
Catalog number: 041239.22
also known as 041239-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9002-89-5
Molecular Formula
(C2H4O)n
SMILES
OC(-*)C-*
MDL Number
MFCD00081922
Viscosity
23.0-27.0 cP (4% solution)
Appearance (Color)
White to off-white
Ash
≤0.5% (Na₂O)
Assay (special)
Methanol: ≤1.0%
Volatiles
≤5.0%

Description

Polyvinyl alcohol is used as a biological reagent to test the dispersion of zinc oxide nanoparticles. It is an effective protein resistant material and is used to stabilize gold colloids of diameter 3-8nm. It is used as an emulsion polymerization aid to make polyvinyl acetate, as a protective colloid, as a film used in the water transfer printing process, as a surfactant for the formation of polymer encapsulated nanobeads, as an embolization agent in medical procedures and as a fixative for specimen collection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text