Thermo Scientific Chemicals
Polyvinyl alcohol, 87-89% hydrolyzed, high molecular weight, Thermo Scientific Chemicals
Catalog number: 041240.14
25 g, Each
Thermo Scientific Chemicals
Polyvinyl alcohol, 87-89% hydrolyzed, high molecular weight, Thermo Scientific Chemicals
Catalog number: 041240.14
25 g, Each
Quantity
Catalog number: 041240.14
also known as 041240-14
Price (USD)
Quantity
-
Have Questions?
Description
Polyvinyl alcohol is used as a biological reagent to test the dispersion of zinc oxide nanoparticles. It is an effective protein resistant material and is used to stabilize gold colloids of diameter 3-8nm. It is used as an emulsion polymerization aid to make polyvinyl acetate, as a protective colloid, as a film used in the water transfer printing process, as a surfactant for the formation of polymer encapsulated nanobeads, as an embolization agent in medical procedures and as a fixative for specimen collection.