Thermo Scientific Chemicals

trans-Dichlorobis(triphenylphosphine)palladium(II), Premion™, 99.95% (metals basis), Pd 14.7% min, Thermo Scientific Chemicals

Catalog number: 041245.06
5 g, Each
Thermo Scientific Chemicals

trans-Dichlorobis(triphenylphosphine)palladium(II), Premion™, 99.95% (metals basis), Pd 14.7% min, Thermo Scientific Chemicals

Catalog number: 041245.06
5 g, Each
Quantity
Catalog number: 041245.06
also known as 041245-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13965-03-2
IUPAC Name
palladium(2+) bis(triphenylphosphane) dichloride
Molecular Formula
C36H30Cl2P2Pd
InChI Key
YNHIGQDRGKUECZ-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Total Metal Impurities
0.05% max., Pd 14.7% min

Description

As a catalyst of choice in hydrogenation, hydrosilation, carbonylation, oxidation, elimination, reduction, allyl aceate isomerization, dimerization of 1,3-dienes, and C-C bond formation such as Negishi coupling, Suzuki coupling, Kumada coupling, Sonogashira coupling, Heck coupling and Sonogashira-Hagihara coupling reactions. Used in the preparation of tetrakis(triphenylphosphine)palladium, condensed heterocycles such as furoquinolines, diphenylacetylenes, and several other compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text