Zinc chloride hydrate, 99.99% (metals basis), Thermo Scientific Chemicals
Zinc chloride hydrate, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc chloride hydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number 041247.18
also known as 041247-18
Price (USD)/ Each
154.65
Online exclusive
172.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
154.65
Online exclusive
172.00 
Save 17.35 (10%)
Add to cart
Zinc chloride hydrate, 99.99% (metals basis), Thermo Scientific Chemicals
Catalog number041247.18
Price (USD)/ Each
154.65
Online exclusive
172.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS29604-34-0
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Zinc(II) chloride hydrate is used as a soldering flux and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is employed as a mordant in printing and dyeing materials, and in the vulcanizing processes of fiber and rubber. It forms a complex with Ruhemann’s purple, which obtained by reacting ninhydrin with amino acids and amines is used in fingerprint detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only