Thermo Scientific Chemicals
Glassy carbon splinter powder, 20-50 micron, type 1, Thermo Scientific Chemicals
Catalog number: 041258.18
50 g, Each
Thermo Scientific Chemicals
Glassy carbon splinter powder, 20-50 micron, type 1, Thermo Scientific Chemicals
Catalog number: 041258.18
50 g, Each
Quantity
Catalog number: 041258.18
also known as 041258-18
Price (USD)
Quantity
-
Have Questions?
Form
Powder, 20-50 micron
Description
Glassy carbon splinter powder is used as such in purifying metals, in electrodes, electrical devices & steel.