Thermo Scientific Chemicals

8-Hydroxyquinoline, ACS, Thermo Scientific Chemicals

Catalog number: 041272.A1
1 kg, Each
Thermo Scientific Chemicals

8-Hydroxyquinoline, ACS, Thermo Scientific Chemicals

Catalog number: 041272.A1
1 kg, Each
Quantity
Catalog number: 041272.A1
also known as 041272-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
148-24-3
IUPAC Name
quinolin-8-ol
Molecular Formula
C9H7NO
InChI Key
MCJGNVYPOGVAJF-UHFFFAOYSA-N
SMILES
OC1=C2N=CC=CC2=CC=C1
Form
Powder or crystals or crystalline powder
Appearance (Color)
White to pale pink to pale brown
Assay from Supplier's CofA
≥99.0% (titration)
Comment
Suitability for magnesium determination: Passes Test
Residue on ignition
≤0.05%

Description

A bacteriostatic and fungistatic agent. Also a chelating agent for metals.8-Hydroxyquinoline is used as an antiseptic and disinfectant. Its alcoholic solution is used in liquid bandages. It is used to prepare tris(8-hydroxyquinolinato)aluminum, which finds application as a component of organic light-emitting diodes (OLED's). It is also used as a reagent and as a stabilizer of hydrogen peroxide in a rocket fuel oxidizer. It is used as a chelating agent and used as a carrier for radio-indium for diagnostic purposes. Its halogenated derivatives are used as anti-infective agents and oral antiamebics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text