Yttria Stabilized Zirconium Oxide Cylindrical Crucibles
Yttria Stabilized Zirconium Oxide Cylindrical Crucibles
Thermo Scientific Chemicals

Yttria Stabilized Zirconium Oxide Cylindrical Crucibles

Have Questions?
Change viewbuttonViewtableView
Diameter (Metric) Outer:
37mm
24mm
45mm
Height (Metric):
View all
72mm
Capacity (Metric):
View all
50 mL
Catalog number 041275.KF
also known as 041275-KF
Price (USD)/ Each
-
Diameter (Metric) Outer:
37mm
Height (Metric):
72mm
Capacity (Metric):
50 mL
Request bulk or custom format
Specifications
MaterialZirconium Oxide
Quantity1 Piece
Capacity (Metric)50 mL
Diameter (Metric) Outer37mm
Height (Metric)72mm
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only