Thermo Scientific Chemicals

Sodium hydroxide monohydrate, 99.996% (metals basis), Thermo Scientific Chemicals

Catalog number: 041281.22
100 g, Each
Thermo Scientific Chemicals

Sodium hydroxide monohydrate, 99.996% (metals basis), Thermo Scientific Chemicals

Catalog number: 041281.22
100 g, Each
Quantity
Catalog number: 041281.22
also known as 041281-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12179-02-1
IUPAC Name
sodium hydrate hydroxide
Molecular Formula
H3NaO2
InChI Key
JBJWASZNUJCEKT-UHFFFAOYSA-M
SMILES
O.[OH-].[Na+]
Total Metal Impurities
0.004% max.

Description

Sodium hydroxide monohydrate is used as an acid neutralizer. It is a strong base widely used in the chemical industry to drive chemical reactions. It finds application as a neutralizing agent in the petroleum refining industry, as paint stripper and an analytical reagent in laboratories to measure the concentration of acids by titration. It is involved in soap production, alumina production by Bayer process, paper industry. It plays an important role as a catalyst for the transesterification of methanol and triglycerides in order to prepare biodiesel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text