Zinc neodecanoate, Zn 17.9-18.2%, Thermo Scientific Chemicals
Zinc neodecanoate, Zn 17.9-18.2%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc neodecanoate, Zn 17.9-18.2%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Catalog number 041312.A7
also known as 041312-A7
Price (USD)/ Each
357.65
Online exclusive
397.00 
Save 39.35 (10%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
357.65
Online exclusive
397.00 
Save 39.35 (10%)
Add to cart
Zinc neodecanoate, Zn 17.9-18.2%, Thermo Scientific Chemicals
Catalog number041312.A7
Price (USD)/ Each
357.65
Online exclusive
397.00 
Save 39.35 (10%)
-
Add to cart
Chemical Identifiers
CAS27253-29-8
IUPAC Namezinc(2+) bis(3,3,5,5-tetramethylhexanoate)
Molecular FormulaC20H38O4Zn
InChI KeyJUODUXKPGONLIM-UHFFFAOYSA-L
SMILES[Zn++].CC(C)(C)CC(C)(C)CC([O-])=O.CC(C)(C)CC(C)(C)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)17.9 - 18.2 % Zn
CommentNon-volatile matter: >/= 92.0 %
Zinc neodecanoate is used as a catalyst, lubricant additive and drier. Further it is used in the production of rubber and plastic products. In addition it serves as a polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only