Thermo Scientific Chemicals
Multi-Element QC-21 Check Standard Solution, Specpure™, Thermo Scientific Chemicals
Catalog number: 041328.AE
100 mL, Each
Thermo Scientific Chemicals
Multi-Element QC-21 Check Standard Solution, Specpure™, Thermo Scientific Chemicals
Catalog number: 041328.AE
100 mL, Each
Quantity
Catalog number: 041328.AE
also known as 041328-AE
Price (USD)
Have Questions?
Concentration
As 100?g/mL ?2%
Concentration
Be 100?g/mL ?2%
Concentration
Ca 100?g/mL ?2%
Concentration
Cd 100?g/mL ?2%
Concentration
Co 100?g/mL ?2%
Description
Multi-Element QC-21 Check Standard Solution is used as a certified reference material or quality control check standard for inductively coupled plasma optical emission spectroscopy (ICP-OES). It is also used in inductively coupled plasma mass spectrometry (ICP-MS), and flame or furnace atomic absorption spectroscopy.