Thermo Scientific Chemicals

Allyl acetate, 97%, Thermo Scientific Chemicals

Catalog number: 041353.AD
50 mL, Each
Thermo Scientific Chemicals

Allyl acetate, 97%, Thermo Scientific Chemicals

Catalog number: 041353.AD
50 mL, Each
Quantity
Catalog number: 041353.AD
also known as 041353-AD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
591-87-7
IUPAC Name
prop-2-en-1-yl acetate
Molecular Formula
C5H8O2
InChI Key
FWZUNOYOVVKUNF-UHFFFAOYSA-N
SMILES
CC(=O)OCC=C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0%

Description

Ally acetate is used in industrial intermediate. Used in synthesis of colophony and bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text