Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis), Thermo Scientific Chemicals
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis), Thermo Scientific Chemicals

Precursor for CVD process
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
0.5 g
2 g
Catalog number 041359.09
also known as 041359-09
Price (USD)/ Each
730.65
Online exclusive
812.00 
Save 81.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
730.65
Online exclusive
812.00 
Save 81.35 (10%)
Add to cart
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II), 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number041359.09
Price (USD)/ Each
730.65
Online exclusive
812.00 
Save 81.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialBis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II)
CAS13986-53-3
Health Hazard 1H317-H341
Health Hazard 2GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(II) is used in nano-material, thin films and ferroelectric metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only