Thermo Scientific Chemicals

Tetramethylammonium hydrogen phthalate, 99+%, Thermo Scientific Chemicals

Catalog number: 041360.22
100 g, Each
Thermo Scientific Chemicals

Tetramethylammonium hydrogen phthalate, 99+%, Thermo Scientific Chemicals

Catalog number: 041360.22
100 g, Each
Quantity
Catalog number: 041360.22
also known as 041360-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79723-02-7
IUPAC Name
tetramethylazanium 2-carboxybenzoate
Molecular Formula
C12H17NO4
InChI Key
KQTFRASJMFIJPC-UHFFFAOYSA-M
SMILES
C[N+](C)(C)C.OC(=O)C1=CC=CC=C1C([O-])=O
Appearance (Color)
White
Assay from Supplier's CofA
>99%
Form
Crystalline powder

Description

Tetramethylammonium hydrogen phthalate is used in electrolyte formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text