Thermo Scientific Chemicals

Xylenol Orange tetrasodium salt, ACS

Catalog number: 041379.04
2 g, Each
Thermo Scientific Chemicals

Xylenol Orange tetrasodium salt, ACS

Catalog number: 041379.04
2 g, Each
Quantity
Catalog number: 041379.04
also known as 041379-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3618-43-7
IUPAC Name
tetrasodium 2-[({5-[3-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-2,2-dioxo-3H-1,2λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxylatomethyl)amino]acetate
Molecular Formula
C31H28N2Na4O13S
InChI Key
FXYMNKLSOQONJB-UHFFFAOYSA-J
SMILES
[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(C2=CC=CC=C2OS1(=O)=O)C1=CC(C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1
Comment
Clarity of solution P.T.
Comment
Suitability for zinc titration P.T.

Description

Xylenol Orange tetrasodium salt is used as a photometric reagent and a metal indicator. It is a versatile indicator for EDTA titrations in acidic range. It is a common organic reagent used as an indicator for metal titrations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text