Thermo Scientific Chemicals

Potassium tert-pentyloxide, 14-18% w/v in cyclohexane, Thermo Scientific Chemicals

Catalog number: 041394.22
100 g, Each
Thermo Scientific Chemicals

Potassium tert-pentyloxide, 14-18% w/v in cyclohexane, Thermo Scientific Chemicals

Catalog number: 041394.22
100 g, Each
Quantity
Catalog number: 041394.22
also known as 041394-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
41233-93-6
IUPAC Name
potassium 2-methylbutan-2-olate
Molecular Formula
C5H11KO
InChI Key
ZRLVQFQTCMUIRM-UHFFFAOYSA-N
SMILES
[K+].CCC(C)(C)[O-]
Form
Liquid
Free base (titration)
≤0.20% (as KOH)
Concentration (weight/weight)
14.0-16.0%
Appearance (Color)
Clear colorless to pale yellow to pale orange

Description

Potassium tert-pentyloxide is used in wide range of applications in organic synthesis, agrochemicals, pharmaceuticals, colorants and aroma chemicals. It is also used in manufacturing detergents and biodiesel. They also act as catalysts in polymerization and isomerizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text