Thermo Scientific Chemicals

Vinylene carbonate, 97+%, stab. with BHT, Thermo Scientific Chemicals

Catalog number: 041400.02
0.5 g, Each
Thermo Scientific Chemicals

Vinylene carbonate, 97+%, stab. with BHT, Thermo Scientific Chemicals

Catalog number: 041400.02
0.5 g, Each
Quantity
Catalog number: 041400.02
also known as 041400-02
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
872-36-6
IUPAC Name
2H-1,3-dioxol-2-one
Molecular Formula
C3H2O3
InChI Key
VAYTZRYEBVHVLE-UHFFFAOYSA-N
SMILES
O=C1OC=CO1
Appearance (Color)
White to pale yellow
Assay from Supplier's CofA
≥97.0% (GC)
Stabilizer
BHT: <2%
Form
Fused or crystalline low melting solid

Description

Vinylene carbonate is used as an additive to electrolyte solutions for anode side Lithium-ion batteries. It also acts as a sealant to seal at least a portion of the silicon-polyvinyl acid interface. It is further used for great improvement of high temperature performance of the battery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text