Vinylene carbonate, 97+%, stab. with BHT, Thermo Scientific Chemicals
Vinylene carbonate, 97+%, stab. with BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vinylene carbonate, 97+%, stab. with BHT, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
0.5 g
2 g
Catalog number 041400.09
Price (USD)/ Each
249.65
Online exclusive
277.00 
Save 27.35 (10%)
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
249.65
Online exclusive
277.00 
Save 27.35 (10%)
Add to cart
Vinylene carbonate, 97+%, stab. with BHT, Thermo Scientific Chemicals
Catalog number041400.09
Price (USD)/ Each
249.65
Online exclusive
277.00 
Save 27.35 (10%)
-
Add to cart
Chemical Identifiers
CAS872-36-6
IUPAC Name2H-1,3-dioxol-2-one
Molecular FormulaC3H2O3
InChI KeyVAYTZRYEBVHVLE-UHFFFAOYSA-N
SMILESO=C1OC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay from Supplier's CofA≥97.0% (GC)
StabilizerBHT: <2%
FormFused or crystalline low melting solid
Vinylene carbonate is used as an additive to electrolyte solutions for anode side Lithium-ion batteries. It also acts as a sealant to seal at least a portion of the silicon-polyvinyl acid interface. It is further used for great improvement of high temperature performance of the battery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only