Cerium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
Cerium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cerium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

For vacuum deposition
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 041408.18
also known as 041408-18
Price (USD)/ Each
310.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
310.00
Add to cart
Cerium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals
Catalog number041408.18
Price (USD)/ Each
310.00
-
Add to cart
Chemical Identifiers
CAS7758-88-5
IUPAC Namecerium(3+) trifluoride
Molecular FormulaCeF3
InChI KeyQCCDYNYSHILRDG-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Ce+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only