Thermo Scientific Chemicals

Gadolinium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 041412.09
10 g, Each
Thermo Scientific Chemicals

Gadolinium(III) fluoride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

Catalog number: 041412.09
10 g, Each
Quantity
Catalog number: 041412.09
also known as 041412-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13765-26-9
IUPAC Name
gadolinium(3+) trifluoride
Molecular Formula
F3Gd
InChI Key
TYIZUJNEZNBXRS-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Gd+3]
Total Rare Earth Oxide Impurities
0.1% max.

Description

Gadolinium(III) fluoride, anhydrous is employed in the synthesis of non-oxide glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text