Yttrium(III) fluoride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Yttrium(III) fluoride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium(III) fluoride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

For vacuum deposition
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Catalog number 041424.18
also known as 041424-18
Price (USD)/ Each
252.00
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
252.00
Add to cart
Yttrium(III) fluoride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number041424.18
Price (USD)/ Each
252.00
-
Add to cart
Chemical Identifiers
CAS13709-49-4
IUPAC Nameyttrium(3+) trifluoride
Molecular FormulaF3Y
InChI KeyRBORBHYCVONNJH-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Yttrium(III) fluoride is used for the production of metallic yttrium, thin films, glasses, electronics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only