Thermo Scientific Chemicals

Yttrium(III) fluoride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 041424.30
250 g, Each
Thermo Scientific Chemicals

Yttrium(III) fluoride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 041424.30
250 g, Each
Quantity
Catalog number: 041424.30
also known as 041424-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13709-49-4
IUPAC Name
yttrium(3+) trifluoride
Molecular Formula
F3Y
InChI Key
RBORBHYCVONNJH-UHFFFAOYSA-K
SMILES
[F-].[F-].[F-].[Y+3]
Total Metal Impurities
0.1% max.

Description

Yttrium(III) fluoride is used for the production of metallic yttrium, thin films, glasses, electronics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text