Silver wire, 0.25mm (0.01in) dia, hard, 99.9% (metals basis), Thermo Scientific Chemicals
Silver wire, 0.25mm (0.01in) dia, hard, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver wire, 0.25mm (0.01in) dia, hard, 99.9% (metals basis), Thermo Scientific Chemicals

Form: Approx. 0.51g/m; Temper: hard
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 m
25 m
500 m
Catalog number 041455.H4
also known as 041455-H4
Price (USD)/ Each
324.00
-
Add to cart
Quantity:
100 m
Request bulk or custom format
Price (USD)/ Each
324.00
Add to cart
Silver wire, 0.25mm (0.01in) dia, hard, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number041455.H4
Price (USD)/ Each
324.00
-
Add to cart
Specifications
Chemical Name or MaterialSilver
ColorSilver
Name NoteHard
CAS7440-22-4
AppearanceSilver-colored wire
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only