Thermo Scientific Chemicals

2-Propanol, anhydrous, 99.5+%, Thermo Scientific Chemicals

Catalog number: 041463.AK
250 mL, Each
Thermo Scientific Chemicals

2-Propanol, anhydrous, 99.5+%, Thermo Scientific Chemicals

Catalog number: 041463.AK
250 mL, Each
Quantity
Catalog number: 041463.AK
also known as 041463-AK
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-63-0
IUPAC Name
propan-2-ol
Molecular Formula
C3H8O
InChI Key
KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES
CC(C)O
Appearance (Color)
Clear, colorless
Form
Liquid
Assay (GC)
≥99.50%
Water Content (Karl Fischer Titration)
100 ppm, max
Residue on Evaporation
5 ppm, max

Description

Solvent for Sol-Gel precursors2-Propanol acts as a solvent for pharmaceuticals, perfumes, gums, resins, alkaloids and essential oils. It is used in cosmetic lotions. Isopropyl alcohol is used as a cleaning agent for keyboards, LCD displays and laptops. It is used as a wetting agent in the fountain solution used in lithographic printing and is often used as a solvent for french polishing shellac, which is used in cabinet making. It acts as a hydride source in the popular Meerwein-Ponndorf-Verley reduction. It is used in huge quantities as a preferred solvent in the manufacture of fine chemicals, drugs, drug intermediates and formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Water <100ppm
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text