Thermo Scientific Chemicals

1-Propanol, anhydrous, 99.9%, Thermo Scientific Chemicals

Catalog number: 041465.K2
1 L, Each
Thermo Scientific Chemicals

1-Propanol, anhydrous, 99.9%, Thermo Scientific Chemicals

Catalog number: 041465.K2
1 L, Each
Quantity
Catalog number: 041465.K2
also known as 041465-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71-23-8
IUPAC Name
propan-1-ol
Molecular Formula
C3H8O
InChI Key
BDERNNFJNOPAEC-UHFFFAOYSA-N
SMILES
CCCO
Appearance (Color)
Colorless
Assay (GC)
≥99.9%
Water Content (Karl Fischer Titration)
50 ppm, max
Residue on Evaporation
3 ppm, max
Form
liquid

Description

Solvent1-Propanol is used as a solvent and an entrainer in azeotropic distillation. It is used as a precursor to prepare propyl acetate, propylamide and propyl halides by reacting with acetic acid, ammonia and halogens respectively. It is actively used as a solvent in inks for flexographic printing, which is a technique used for printing. It is added to cleaners, floor polishes and metal degreasing fluids and is used as an additional solvent in the manufacture of adhesives. It enhances the drying characteristics of alkyd resins, electrodeposition paints and baking finishes in coating industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text