Thermo Scientific Chemicals

tert-Butyl alcohol, anhydrous, 99.5%, Thermo Scientific Chemicals

Catalog number: 041470.K2
1 L, Each
Thermo Scientific Chemicals

tert-Butyl alcohol, anhydrous, 99.5%, Thermo Scientific Chemicals

Catalog number: 041470.K2
1 L, Each
Quantity
Catalog number: 041470.K2
also known as 041470-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75-65-0
IUPAC Name
2-methylpropan-2-ol
Molecular Formula
C4H10O
InChI Key
DKGAVHZHDRPRBM-UHFFFAOYSA-N
SMILES
CC(C)(C)O
Appearance (Color)
Clear, colorless
Form
liquid ( when melted)
Assay (GC)
>/=99.5%
Water Content (Karl Fischer Titration)
100 ppm, max
Residue on Evaporation
5 ppm, max

Description

Solvent in Sol-Gel precursor applications.tert-Butyl alcohol is used as a solvent and an ethanol denaturant. It is an active ingredient paint remover. It acts as an intermediate in the production of methyl tertiary butyl ether and ethyl tertiary butyl ether by reaction with methanol and ethanol. It is also used in the preparation of tert-butyl chloride, tert-butyl hydroperoxide, isobutylene and methyl methacrylate. It serves as a fuel additive as well as a gasoline octane booster.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text