tert-Butyl alcohol, anhydrous, 99.5%, Thermo Scientific Chemicals
tert-Butyl alcohol, anhydrous, 99.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl alcohol, anhydrous, 99.5%, Thermo Scientific Chemicals

Solvent in Sol-Gel precursor applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
250 mL
1 L
Catalog number 041470.K7
also known as 041470-K7
Price (USD)/ Each
372.00
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
372.00
Add to cart
tert-Butyl alcohol, anhydrous, 99.5%, Thermo Scientific Chemicals
Catalog number041470.K7
Price (USD)/ Each
372.00
-
Add to cart
Chemical Identifiers
CAS75-65-0
IUPAC Name2-methylpropan-2-ol
Molecular FormulaC4H10O
InChI KeyDKGAVHZHDRPRBM-UHFFFAOYSA-N
SMILESCC(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear, colorless
Formliquid ( when melted)
Assay (GC)>/=99.5%
Water Content (Karl Fischer Titration)100 ppm, max
Residue on Evaporation5 ppm, max
Solvent in Sol-Gel precursor applications.tert-Butyl alcohol is used as a solvent and an ethanol denaturant. It is an active ingredient paint remover. It acts as an intermediate in the production of methyl tertiary butyl ether and ethyl tertiary butyl ether by reaction with methanol and ethanol. It is also used in the preparation of tert-butyl chloride, tert-butyl hydroperoxide, isobutylene and methyl methacrylate. It serves as a fuel additive as well as a gasoline octane booster.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only