4,4'-Diaminobiphenyl-2,2'-disulfonic acid hydrate, cont. up to 30% water, Thermo Scientific Chemicals
4,4'-Diaminobiphenyl-2,2'-disulfonic acid hydrate, cont. up to 30% water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,4'-Diaminobiphenyl-2,2'-disulfonic acid hydrate, cont. up to 30% water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041472.22
also known as 041472-22
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
Add to cart
4,4'-Diaminobiphenyl-2,2'-disulfonic acid hydrate, cont. up to 30% water, Thermo Scientific Chemicals
Catalog number041472.22
Price (USD)/ Each
154.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
182.00 
Save 27.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name4,4'-diamino-[1,1'-biphenyl]-2,2'-disulfonic acid
Molecular FormulaC12H12N2O6S2
InChI KeyMBJAPGAZEWPEFB-UHFFFAOYSA-N
SMILESNC1=CC=C(C2=CC=C(N)C=C2S(O)(=O)=O)C(=C1)S(O)(=O)=O
Molecular Weight (g/mol)344.36
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to pink to reddish purple
FormPowder or crystals or crystalline powder
Assay (unspecified)≥70.0%
Loss on Drying≤30.0%
4,4'-Diaminobiphenyl-2,2'-disulfonic acid hydrate is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only