Thermo Scientific Chemicals

Trifluoroiodomethane, 97+%, Thermo Scientific Chemicals

Catalog number: 041479.22
100 g, Each
Thermo Scientific Chemicals

Trifluoroiodomethane, 97+%, Thermo Scientific Chemicals

Catalog number: 041479.22
100 g, Each
Quantity
Catalog number: 041479.22
also known as 041479-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2314-97-8
IUPAC Name
trifluoro(iodo)methane
Molecular Formula
CF3I
InChI Key
VPAYJEUHKVESSD-UHFFFAOYSA-N
SMILES
FC(F)(F)I
Assay (unspecified)
> 97.0%

Description

Trifluoroiodomethane is used as a gaseous fire suppression flooding agent for in-flight aircraft and electronic equipment fires. It is also an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It is involved in the rhodium-catalyzed alfa-trifluoromethylation of alfa,beta-unsaturatedketones. It plays an important role as catalyst in the enantioselective alfa -trifluoromethylation of aldehydes through photoredox organocatalysis using a readily available iridium photocatalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text