Thermo Scientific Chemicals

Chloromethanesulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: 041486.04
2 g, Each
Thermo Scientific Chemicals

Chloromethanesulfonyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: 041486.04
2 g, Each
Quantity
Catalog number: 041486.04
also known as 041486-04
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3518-65-8
IUPAC Name
chloromethanesulfonyl chloride
Molecular Formula
CH2Cl2O2S
InChI Key
KQDDQXNVESLJNO-UHFFFAOYSA-N
SMILES
ClCS(Cl)(=O)=O
Form
Liquid
Assay (Titration)
≥94.0%
Appearance (Color)
Clear Colorless to yellow or pale brown

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff. It is synthesis of chloromethanesulfonates and chloromethanesulfonamides. It is also used for conversion of alcohols into the inverted acetates, azides and nitriles, and rearrangement of carbon-carbon and carbon-oxygen bonds under mild conditions, and in electrophilic and radical reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text