Yttrium aluminum oxide, 99.9% (REO), Thermo Scientific Chemicals
Yttrium aluminum oxide, 99.9% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Yttrium aluminum oxide, 99.9% (REO), Thermo Scientific Chemicals

Rare earth element doped yttrium aluminum oxide is used in solid state lasers
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 041491.14
also known as 041491-14
Price (USD)/ Each
194.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
194.00
Add to cart
Yttrium aluminum oxide, 99.9% (REO), Thermo Scientific Chemicals
Catalog number041491.14
Price (USD)/ Each
194.00
-
Add to cart
Chemical Identifiers
CAS12005-21-9
IUPAC Namealuminium(3+) yttrium(3+) trioxidandiide
Molecular FormulaAlO3Y
InChI KeyJNDMLEXHDPKVFC-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Rare earth element doped yttrium aluminum oxide is used in solid state lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only