Thermo Scientific Chemicals

Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, soln. in vinyl terminated polydimethylsiloxane, Thermo Scientific Chemicals

Catalog number: 041508.14
25 g, Each
Thermo Scientific Chemicals

Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, soln. in vinyl terminated polydimethylsiloxane, Thermo Scientific Chemicals

Catalog number: 041508.14
25 g, Each
Quantity
Catalog number: 041508.14
also known as 041508-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68478-92-2
Molecular Formula
C8 H18 O Pt Si2
Molecular Weight (g/mol)
474.68
MDL Number
MFCD00151662
Appearance (Color)
Orange
Elemental Analysis
% Pt 3.0 - 3.5%
Form
Hazy liquid

Description

Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst.Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, soln. in vinyl terminated polydimethylsiloxane is used as a catalyst in hydrosilylation reaction for the preparation of trans-beta-silanes and in reductive amination reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text