Cadmium sulfide, 99.99% (metals basis), Thermo Scientific Chemicals
Cadmium sulfide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium sulfide, 99.99% (metals basis), Thermo Scientific Chemicals

Vacuum deposition
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041511.14
also known as 041511-14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1306-23-6
IUPAC Namecadmium(2+) sulfanediide
Molecular FormulaCdS
InChI KeyFRLJSGOEGLARCA-UHFFFAOYSA-N
SMILES[S--].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only