Di-n-octyl phthalate, 98%, Thermo Scientific Chemicals
Di-n-octyl phthalate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Di-n-octyl phthalate, 98%, Thermo Scientific Chemicals

Plasticizer
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Catalog number 041522.AP
also known as 041522-AP
Price (USD)/ Each
472.65
Online exclusive
525.00 
Save 52.35 (10%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
472.65
Online exclusive
525.00 
Save 52.35 (10%)
Add to cart
Di-n-octyl phthalate, 98%, Thermo Scientific Chemicals
Catalog number041522.AP
Price (USD)/ Each
472.65
Online exclusive
525.00 
Save 52.35 (10%)
-
Add to cart
Specifications
Chemical Name or MaterialDi-n-octyl phthalate
CAS117-84-0
Melting Point-25°C
ColorColorless to Yellow
Recommended StorageAmbient temperatures
View more
Di-n-octyl phthalate is used as a plasticizer for many resins and elastomers. It acts as an additive. It is also used for medical tubing and blood storage bags, wire and cables, carpet back coating, floor tile and in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only