Thermo Scientific Chemicals

Sodium arsenite, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 041533.AP
500 mL, Each
Thermo Scientific Chemicals

Sodium arsenite, 0.1N Standardized Solution, Thermo Scientific Chemicals

Catalog number: 041533.AP
500 mL, Each
Quantity
Catalog number: 041533.AP
also known as 041533-AP
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7784-46-5
IUPAC Name
sodium oxoarsinite
Molecular Formula
AsNaO2
InChI Key
PTLRDCMBXHILCL-UHFFFAOYSA-M
SMILES
[Na+].[O-][As]=O
Concentration
Sodium arsenite 0.1N

Description

Sodium arsenite is used as hide preservative, antiseptic, dyeing and soaps. It is used as a stressor to induce production of heat shock proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text