Thermo Scientific™

3,4-Dihydroxy-DL-phenylalanine, 98%

Catalog number: 041535.06
5 g, Each
Thermo Scientific™

3,4-Dihydroxy-DL-phenylalanine, 98%

Catalog number: 041535.06
5 g, Each
Quantity
Catalog number: 041535.06
also known as 041535-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63-84-3
IUPAC Name
2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
Molecular Formula
C9H11NO4
InChI Key
WTDRDQBEARUVNC-UHFFFAOYNA-N
SMILES
NC(CC1=CC=C(O)C(O)=C1)C(O)=O
Appearance (Color)
White to brown to gray
Form
Powder
Assay (HPLC)
≥97.5%

Description

3,4-Dihydroxy-DL-phenylalanine is used as a dopamine precursor. It is also useful in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text