Antimony(III) chloride, 99.9% (metals basis), Thermo Scientific Chemicals
Antimony(III) chloride, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimony(III) chloride, 99.9% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 041537.22
also known as 041537-22
Price (USD)/ Each
165.65
Online exclusive
184.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
165.65
Online exclusive
184.00 
Save 18.35 (10%)
Add to cart
Antimony(III) chloride, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number041537.22
Price (USD)/ Each
165.65
Online exclusive
184.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10025-91-9
IUPAC Nameantimony(3+) trichloride
Molecular FormulaCl3Sb
InChI KeyFAPDDOBMIUGHIN-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Sb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols. It is a complexing agent used in the colorimetric Carr-Price test for detecting carotenoids including vitamin A. It is employed as a mordant and a fabric flame retardant. It is used as a catalyst for chlorinationn and polymerization reactions. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine. It is used as an analytical reagent for chloral and aromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only