Thermo Scientific Chemicals

Potash, sulfurated, S 11.8% min as sulfide, Thermo Scientific Chemicals

Catalog number: 041550.36
500 g, Each
Thermo Scientific Chemicals

Potash, sulfurated, S 11.8% min as sulfide, Thermo Scientific Chemicals

Catalog number: 041550.36
500 g, Each
Quantity
Catalog number: 041550.36
also known as 041550-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
39365-88-3
IUPAC Name
tetrapotassium 1-oxo-1λ⁶-disulfene-1,1-bis(olate) sulfanediide
Molecular Formula
K4O3S3
InChI Key
YFKFGPGTVQHNEF-UHFFFAOYSA-L
SMILES
[S--].[K+].[K+].[K+].[K+].[O-]S([O-])(=O)=S
Assay (unspecified)
≥11.8% (Sulfides)
Form
Granules or chunks
Appearance (Color)
Green to yellow to brown to grey

Description

Potash is used in black/brown patina (copper, brass, bronze). Its use include decorative color effects on copper, brass, bronze and nickel. Sulfurated potash has been used in the treatment of acne and other skin disorders usually in the form of a lotion with zinc sulfate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text