Thermo Scientific Chemicals

Calcium boride, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 041551.14
25 g, Each
Thermo Scientific Chemicals

Calcium boride, 99+% (metals basis), Thermo Scientific Chemicals

Catalog number: 041551.14
25 g, Each
Quantity
Catalog number: 041551.14
also known as 041551-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12007-99-7
IUPAC Name
calcium pentacyclo[3.1.0.0¹,³.0²,⁴.0⁴,⁶]hexaborane-1,4-diuide
Molecular Formula
B6Ca
InChI Key
AWVIDOFQNLOVRC-UHFFFAOYSA-N
SMILES
[Ca++].B12B3[B-]11B4B1[B-]234
Total Metal Impurities
1% max.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text