Thermo Scientific Chemicals

Polyethylene glycol monomethylether, 350, Thermo Scientific Chemicals

Catalog number: 041560.22
100 g, Each
Thermo Scientific Chemicals

Polyethylene glycol monomethylether, 350, Thermo Scientific Chemicals

Catalog number: 041560.22
100 g, Each
Quantity
Catalog number: 041560.22
also known as 041560-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-74-4
Molecular Formula
(C2H4O)nCH4O
InChI Key
XNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILES
COCCO
MDL Number
MFCD00084416
Molecular Weight
325 to 375 D
pH
4.5 to 7.5 ( 5% aq sol at 25?C)
Residual water
0.5% max

Description

Polyethylene glycol monomethylether is used as enteric release coatings. It is also used for a series of polycarboxylate water reducing agent. It acts as a solvent for brake fluids. Further, it is used in the building materials industry and raw material for cement water reducing agent and strengthening agent. In addition to this, it is used in surfactants, polyester and polyurethane based paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text