Polyethylene glycol monomethylether, 750, Thermo Scientific Chemicals
Polyethylene glycol monomethylether, 750, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyethylene glycol monomethylether, 750, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 041562.22
also known as 041562-22
Price (USD)/ Each
29.65
Online exclusive
32.70 
Save 3.05 (9%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
29.65
Online exclusive
32.70 
Save 3.05 (9%)
Add to cart
Polyethylene glycol monomethylether, 750, Thermo Scientific Chemicals
Catalog number041562.22
Price (USD)/ Each
29.65
Online exclusive
32.70 
Save 3.05 (9%)
-
Add to cart
Chemical Identifiers
CAS9004-74-4
Molecular Formula(C2H4O)nCH4O
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
MDL NumberMFCD00084416
SpecificationsSpecification SheetSpecification Sheet
Residual water3 % , max
Molecular Weight715 to 785
pH4.5 to 7.5 ( 5% aq. soln., 25 deg C)
FormFused mass/ clear liquid as a melt
Polyethylene glycol monomethylether is used as a solvent, excipient, surfactant and dispersing agent. It is also used as a wetting agent and viscosity modifier. It finds application in the cosmetic and pharmaceutical industry due its low toxicity, lubricating property and solubility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only