Polyethylene glycol monomethylether, 1,900, Thermo Scientific Chemicals
Polyethylene glycol monomethylether, 1,900, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyethylene glycol monomethylether, 1,900, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 041563.36
also known as 041563-36
Price (USD)/ Each
201.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
201.00
Add to cart
Polyethylene glycol monomethylether, 1,900, Thermo Scientific Chemicals
Catalog number041563.36
Price (USD)/ Each
201.00
-
Add to cart
Chemical Identifiers
CAS9004-74-4
Molecular Formula(C2H4O)nCH4O
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
MDL NumberMFCD00084416
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)white
Formflakes or powder
Assay (unspecified)2.0% max, Volatile content
Polyethylene glycol monomethylether is used in the introduction of hydrophilic sites into polymers. It is also used to stabilize polymer emulsions. Further, it is employed in the synthesis of comb polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only