Cadmium acetate dihydrate, 98%, Thermo Scientific Chemicals
Cadmium acetate dihydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium acetate dihydrate, 98%, Thermo Scientific Chemicals

In cadium electroplating
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 041581.22
also known as 041581-22
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
60.70 
Save 9.05 (15%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
60.70 
Save 9.05 (15%)
Add to cart
Cadmium acetate dihydrate, 98%, Thermo Scientific Chemicals
Catalog number041581.22
Price (USD)/ Each
51.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
60.70 
Save 9.05 (15%)
-
Add to cart
Chemical Identifiers
CAS5743-04-4
IUPAC Namecadmium(2+) diacetate dihydrate
Molecular FormulaC4H10CdO6
InChI KeyAUIZLSZEDUYGDE-UHFFFAOYSA-L
SMILESO.O.[Cd++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
Cadmium acetate dihydrate is in the porcelain production for the production of iridescent used effects. Moreover, it is in the analytical chemistry, a reagent for the detection of sulfur, selenium and tellurium. It is also used as chemical and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only