Thermo Scientific Chemicals

Potassium phosphate monohydrate, 95%, Thermo Scientific Chemicals

Catalog number: 041586.A3
2 kg, Each
Thermo Scientific Chemicals

Potassium phosphate monohydrate, 95%, Thermo Scientific Chemicals

Catalog number: 041586.A3
2 kg, Each
Quantity
Catalog number: 041586.A3
also known as 041586-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27176-10-9
IUPAC Name
tripotassium hydrate phosphate
Molecular Formula
H2K3O5P
InChI Key
RMNIZOOYFMNEJJ-UHFFFAOYSA-K
SMILES
O.[K+].[K+].[K+].[O-]P([O-])([O-])=O
Assay (unspecified)
94.1% min

Description

Potassium phosphate monohydrate acts as a reagent and buffer used in molecular biology, biochemistry, and chromatography. It is utilized to study the effects of freezing and thawing of proteins sensitive to conformational changes. It is also used in the extraction of keratohyalin protein from bovine tissue. Further, it is used as a food additive, emulsifier, foaming agent and fertilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text