Calcium hydride, 92% min, Thermo Scientific Chemicals
Calcium hydride, 92% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium hydride, 92% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 041590.36
also known as 041590-36
Price (USD)/ Each
371.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
371.00
Add to cart
Calcium hydride, 92% min, Thermo Scientific Chemicals
Catalog number041590.36
Price (USD)/ Each
371.00
-
Add to cart
Chemical Identifiers
CAS7789-78-8
IUPAC Namecalcium
Molecular FormulaCa
InChI KeyOYPRJOBELJOOCE-UHFFFAOYSA-N
SMILES[Ca]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)92% min
Calcium hydride is used as an efficient drying agent for aprotic base-stable solvents like ethers and tertiary amines. It is a useful dehydrating agent in a synthesis of aldehyde enamines in high yield and purity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only