USA Standard Sieve, 635 Mesh
Product Image
Thermo Scientific Chemicals
USA Standard Sieve, 635 Mesh
Brass frame and Stainless-steel mesh sieves
Have Questions?
Change viewbuttonViewtableView
Diameter (Metric) Outer:
30.4 cm
20.3 cm
7.62 cm
Catalog number 041593.NK
also known as 041593-NK
Price (USD)/ Each
1,304.00
-
Add to cart
Diameter (Metric) Outer:
30.4 cm
Request bulk or custom format
Price (USD)/ Each
1,304.00
Add to cart
USA Standard Sieve, 635 Mesh
Catalog number041593.NK
Price (USD)/ Each
1,304.00
-
Add to cart
Specifications
Description635 Mesh
Depth (English)3 in.
Depth (Metric)7.62 cm
TypeSieve
Certifications/ComplianceASTM E11
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Brass frame, stainless steel mesh sieves meet ASTM E11 requirements and all current amendments
  • Sieves are full height
Sieve Depths
  • 3 in. diameter sieves are 1 in. deep
  • 8 in. diameter sieves are 2 in. deep
  • 12 in.diameter sieves are 3 in. deep
RUO – Research Use Only