Lithium hydride, 99.4% (metals basis), Thermo Scientific Chemicals
Lithium hydride, 99.4% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium hydride, 99.4% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 041596.14
also known as 041596-14
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
Add to cart
Lithium hydride, 99.4% (metals basis), Thermo Scientific Chemicals
Catalog number041596.14
Price (USD)/ Each
109.65
Online exclusive
122.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7580-67-8
IUPAC Namelithium(1+) hydride
Molecular FormulaHLi
InChI KeySRTHRWZAMDZJOS-UHFFFAOYSA-N
SMILES[H-].[Li+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.6% max.
Lithium hydride is used as a precursor in the production of a variety of reagents in the synthesis, especially silane by reacting with silicon tetrachloride through Sundermeyer process and lithium aluminum hydride and lithium borohydride. It finds application in nuclear reactors as a shielding material. In addition, it is a hydrogen source or reducing agent to prepare other hydrides amides and 2H isotopic compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only