Thermo Scientific Chemicals

Silver powder, spherical, -635 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 041600.30
250 g, Each
Thermo Scientific Chemicals

Silver powder, spherical, -635 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 041600.30
250 g, Each
Quantity
Catalog number: 041600.30
also known as 041600-30
Price (EUR)
Quantity
-

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute aquatic toxicity
    Category 1
    Chronic aquatic toxicity
    Category 1
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H410 - Very toxic to aquatic life with long lasting effects

    Precautionary Statements

    P273 - Avoid release to the environment

    P391 - Collect spillage

    P501 - Dispose of contents/ container to an approved waste disposal plant